Assay Details
| Gene Symbol: Gpx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105738.1 | NP_001099208.1 | 115 | 84 | ||
| GenBank mRNA | X62404.1 | - | 160 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpx5
Genomic Map