Assay Details
| Gene Symbol: Pram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241132.2 | XP_006241194.1 | 6 - 7 | 2082 | 122 | |
| XM_008765204.1 | XP_008763426.1 | 6 - 7 | 2082 | 122 | ||
More Information
Set Membership: |
|

Pram1
Genomic Map