Assay Details
Gene Symbol: March2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241126.2 | XP_006241188.1 | 1 - 2 | 140 | 137 | |
XM_006241127.2 | XP_006241189.1 | 1 - 2 | 140 | 137 | ||
XM_008765201.1 | XP_008763423.1 | 1 - 2 | 140 | 137 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006241126.2, XM_006241127.2, XM_008765201.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
