Assay Details
Gene Symbol: March2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034108.1 | NP_001029280.1 | 2 - 3 | 258 | 58 | |
XM_006241126.2 | XP_006241188.1 | 2 - 3 | 373 | 58 | ||
XM_006241127.2 | XP_006241189.1 | 2 - 3 | 373 | 58 | ||
XM_008765201.1 | XP_008763423.1 | 2 - 3 | 373 | 58 | ||
GenBank mRNA | AB048838.1 | - | 1 - 2 | 222 | 58 | |
AI578941.1 | - | 2 - 3 | 258 | 58 | ||
BC088286.1 | - | 1 - 2 | 217 | 7383057 | 58 | |
FQ226318.1 | - | 1 - 2 | 79 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
