Assay Details
Gene Symbol: Ctsz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183330.1 | NP_899159.1 | 5 - 6 | 887 | 83 | |
GenBank mRNA | AB023781.2 | - | 5 - 6 | 887 | 83 | |
BC091110.1 | - | 5 - 6 | 821 | 7379777 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
