Assay Details
Gene Symbol: Spo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108964.1 | NP_001102434.1 | 7 - 8 | 665 | 64 | |
XM_006235698.2 | XP_006235760.1 | 8 - 9 | 825 | 64 | ||
XM_006235701.2 | XP_006235763.1 | 8 - 9 | 825 | 64 | ||
XM_008762524.1 | XP_008760746.1 | 8 - 9 | 825 | 64 | ||
XM_008762525.1 | XP_008760747.1 | 8 - 9 | 1513 | 64 | ||
XM_008762527.1 | XP_008760749.1 | 8 - 9 | 1059 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
