Assay Details
| Gene Symbol: Abcb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133401.1 | NP_596892.1 | 9 - 10 | 1054 | 89 | |
| XM_006235994.2 | XP_006236056.1 | 11 - 12 | 1879 | 89 | ||
| GenBank mRNA | AF257746.1 | - | 9 - 10 | 1054 | 89 | |
| AF286167.2 | - | 8 - 9 | 969 | 89 | ||
| AY582535.1 | - | 8 - 9 | 969 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb1a
Genomic Map