Assay Details
| Gene Symbol: Esco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126299.2 | NP_001119771.2 | 1 - 2 | 1998 | 92 | |
| XM_006254412.2 | XP_006254474.1 | 3 - 4 | 2611 | 92 | ||
| XM_006254413.2 | XP_006254475.1 | 3 - 4 | 2421 | 92 | ||
| XM_006254414.2 | XP_006254476.1 | 3 - 4 | 2608 | 92 | ||
| XM_006254415.2 | XP_006254477.1 | 3 - 4 | 2774 | 92 | ||
| GenBank mRNA | BC166441.1 | - | 1 - 2 | 1998 | 7461161 | 92 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Esco1
Genomic Map