Assay Details
| Gene Symbol: Myl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109484.1 | NP_001102954.1 | 1 - 2 | 70 | 82 | |
| XM_008765039.1 | XP_008763261.1 | 1 - 2 | 64 | 82 | ||
| XM_008765040.1 | XP_008763262.1 | 1 - 2 | 64 | 82 | ||
| GenBank mRNA | BC166586.1 | - | 1 - 2 | 35 | 7934414 | 82 |
| FQ217952.1 | - | 1 - 2 | 43 | 82 | ||
| FQ221005.1 | - | 1 - 2 | 41 | 82 | ||
| FQ221192.1 | - | 1 - 2 | 37 | 82 | ||
| FQ221499.1 | - | 1 - 2 | 39 | 82 | ||
| FQ221709.1 | - | 1 - 2 | 71 | 82 | ||
| FQ221718.1 | - | 1 - 2 | 42 | 82 | ||
| FQ221751.1 | - | 1 - 2 | 38 | 82 | ||
| FQ221963.1 | - | 1 - 2 | 39 | 82 | ||
| FQ222160.1 | - | 1 - 2 | 40 | 82 | ||
| FQ222384.1 | - | 1 - 2 | 42 | 82 | ||
| FQ223317.1 | - | 1 - 2 | 45 | 82 | ||
| FQ228533.1 | - | 1 - 2 | 42 | 82 | ||
| Gene Symbol: Myl6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109486.1 | NP_001102956.1 | 1 - 2 | 70 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC166586.1, NM_001109484.1, NM_001109486.1, XM_008765039.1, XM_008765040.1 transcripts.
Set Membership: |
|

Myl6
Genomic Map