Assay Details
Gene Symbol: Srpk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_184045.2 | NP_908934.1 | 10 - 11 | 1164 | 84 | |
XM_006229558.2 | XP_006229620.1 | 10 - 11 | 1299 | 84 | ||
XM_006229559.2 | XP_006229621.1 | 10 - 11 | 1295 | 84 | ||
XM_006229560.2 | XP_006229622.1 | 10 - 11 | 1290 | 84 | ||
XM_006229561.2 | XP_006229623.1 | 10 - 11 | 1287 | 84 | ||
XM_008773617.1 | XP_008771839.1 | 8 - 9 | 1101 | 84 | ||
XM_008773618.1 | XP_008771840.1 | 8 - 9 | 1091 | 84 | ||
GenBank mRNA | AY346151.1 | - | 10 - 11 | 1168 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
