Assay Details
| Gene Symbol: Naa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135839.1 | NP_001129311.1 | 1 - 2 | 162 | 60 | |
| XM_006229592.2 | XP_006229654.1 | 1 - 2 | 271 | 60 | ||
| GenBank mRNA | FQ235221.1 | - | 1 - 2 | 106 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001135839.1, XM_006229592.2 transcripts.
Set Membership: |
|

Naa10
Genomic Map