Assay Details
Gene Symbol: Gmnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106112.1 | NP_001099582.1 | 2 - 3 | 393 | 80 | |
XM_006253927.2 | XP_006253989.1 | 2 - 3 | 178 | 80 | ||
XM_006253928.2 | XP_006253990.1 | 2 - 3 | 116 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
