Assay Details
| Gene Symbol: Tdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034947.1 | NP_001030119.1 | 1 - 2 | 250 | 70 | |
| GenBank mRNA | BC101920.1 | - | 1 - 2 | 250 | 7937874 | 70 |
More Information
Updated Mapping Information
Set Membership: |
|

Tdp2
Genomic Map