Assay Details
| Gene Symbol: Ebpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108381.1 | NP_001101851.1 | 2 - 3 | 249 | 119 | |
| GenBank mRNA | FQ213140.1 | - | 2 - 3 | 255 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ebpl
Genomic Map