Assay Details
| Gene Symbol: C2cd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_576426.5 | XP_576426.2 | 1 - 2 | 3218 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_576426.5 transcripts.
Set Membership: |
|

C2cd4b
Genomic Map