Assay Details
| Gene Symbol: Myzap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014211.1 | NP_001014233.1 | 1 - 2 | 83 | 69 | |
| XM_008766312.1 | XP_008764534.1 | 1 - 2 | 191 | 69 | ||
| GenBank mRNA | BC089899.1 | - | 1 - 2 | 83 | 7382871 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|

Myzap
Genomic Map