Assay Details
| Gene Symbol: Rsl24d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014212.1 | NP_001014234.1 | 1 - 2 | 135 | 95 | |
| GenBank mRNA | BC062237.1 | - | 1 - 2 | 135 | 6919069 | 95 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rsl24d1
Genomic Map