Assay Details
Gene Symbol: Rps27l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276477.1 | NP_001263406.1 | 4 - 5 | 509 | 137 | |
GenBank mRNA | CB712651.1 | - | 3 - 4 | 259 | 137 | |
FQ109437.1 | - | 4 - 5 | 508 | 137 | ||
FQ209888.1 | - | 3 - 4 | 306 | 137 | ||
X59375.1 | - | 3 - 4 | 307 | 137 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
