Assay Details
| Gene Symbol: Ndufa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001076462.4 | XP_001076462.2 | 3 - 4 | 244 | 83 | |
| GenBank mRNA | BC157820.1 | - | 2 - 3 | 160 | 83 | |
| FQ223729.1 | - | 3 - 4 | 171 | 83 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ndufa3
Genomic Map