Assay Details
| Gene Symbol: Oscar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184973.1 | NP_001171902.1 | 2 - 3 | 146 | 92 | |
| XM_006228020.2 | XP_006228082.1 | 2 - 3 | 367 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Oscar
Genomic Map