Assay Details
| Gene Symbol: Oscar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184973.1 | NP_001171902.1 | 4 - 5 | 724 | 121 | |
| XM_006228020.2 | XP_006228082.1 | 4 - 5 | 945 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Oscar
Genomic Map