Assay Details
| Gene Symbol: Lilrb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031713.1 | NP_113901.2 | 6 - 7 | 1136 | 70 | |
| GenBank mRNA | AF082534.1 | - | 6 - 7 | 1136 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lilrb3
Genomic Map