Assay Details
Gene Symbol: Gprasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773392.1 | XP_008771614.1 | 2 - 3 | 200 | 63 | |
XM_008773394.1 | XP_008771616.1 | 2 - 3 | 200 | 63 | ||
XM_008773395.1 | XP_008771617.1 | 2 - 3 | 200 | 63 | ||
XM_008773397.1 | XP_008771619.1 | 2 - 3 | 200 | 63 | ||
XM_008773398.1 | XP_008771620.1 | 2 - 3 | 200 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008773392.1, XM_008773394.1, XM_008773395.1, XM_008773397.1, XM_008773398.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
