Assay Details
| Gene Symbol: LOC102554842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772258.1 | XP_008770480.1 | 2 - 3 | 381 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102554842| Gene Symbol: LOC102554842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772258.1 | XP_008770480.1 | 2 - 3 | 381 | 67 | |
Set Membership: |
|