Assay Details
| Gene Symbol: Arl2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255120.2 | XP_006255182.1 | 1 - 2 | 266 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006255120.2 transcripts.
Set Membership: |
|

Arl2bp
Genomic Map