Assay Details
| Gene Symbol: RT1-S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008886.2 | NP_001008886.2 | 2 - 3 | 374 | 68 | |
| XM_006255917.2 | XP_006255979.1 | 2 - 3 | 484 | 68 | ||
| GenBank mRNA | AF029241.1 | - | 2 - 3 | 299 | 68 | |
| AJ243973.1 | - | 1 - 2 | 271 | 68 | ||
| AJ243975.1 | - | 1 - 2 | 271 | 68 | ||
| AJ243976.1 | - | 1 - 2 | 271 | 68 | ||
| CK599963.1 | - | 2 - 3 | 374 | 68 | ||
| FQ209394.1 | - | 2 - 3 | 363 | 68 | ||
| FQ232669.1 | - | 2 - 3 | 362 | 68 | ||
| X16979.1 | - | 1 - 2 | 94 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RT1-S3
Genomic Map