Assay Details
Gene Symbol: RT1-S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008886.2 | NP_001008886.2 | 6 - 7 | 1095 | 104 | |
GenBank mRNA | AF029241.1 | - | 6 - 7 | 1020 | 104 | |
AJ243976.1 | - | 5 - 6 | 992 | 104 | ||
FQ209394.1 | - | 6 - 7 | 1085 | 104 | ||
FQ232669.1 | - | 6 - 7 | 1086 | 104 | ||
X16979.1 | - | 5 - 6 | 815 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
