Assay Details
Gene Symbol: RT1-S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008886.2 | NP_001008886.2 | 7 - 8 | 1146 | 98 | |
GenBank mRNA | AF029241.1 | - | 7 - 8 | 1071 | 98 | |
FQ209394.1 | - | 7 - 8 | 1136 | 98 | ||
FQ232669.1 | - | 7 - 8 | 1137 | 98 | ||
X16979.1 | - | 6 - 7 | 866 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
