Assay Details
| Gene Symbol: RT1-T24-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166403.1 | NP_001159875.1 | 5 - 6 | 1036 | 64 | |
| XM_008772749.1 | XP_008770971.1 | 4 - 5 | 1519 | 64 | ||
| GenBank mRNA | FQ230636.1 | - | 4 - 5 | 795 | 64 | |
| FQ234092.1 | - | 4 - 5 | 794 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RT1-T24-3
Genomic Map