Assay Details
Gene Symbol: RT1-T24-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166403.1 | NP_001159875.1 | 4 - 5 | 900 | 66 | |
XM_008772752.1 | XP_008770974.1 | 4 - 5 | 1276 | 66 | ||
GenBank mRNA | FQ234092.1 | - | 3 - 4 | 658 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
