Assay Details
| Gene Symbol: Hnrnpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC088150.1 | - | 10 - 11 | 1096 | 7374689 | 104 |
| FQ234419.1 | - | 10 - 11 | 1112 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hnrnpa1
Genomic Map