Assay Details
| Gene Symbol: Hnrnpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017248.1 | NP_058944.1 | 1 - 2 | 48 | 132 | |
| XM_006242384.1 | XP_006242446.1 | 1 - 2 | 386 | 132 | ||
| GenBank mRNA | BC062235.1 | - | 1 - 2 | 85 | 6920834 | 132 |
| BC088150.1 | - | 1 - 2 | 65 | 7374689 | 132 | |
| FQ234419.1 | - | 1 - 2 | 81 | 132 | ||
| M12156.1 | - | 1 - 2 | 48 | 132 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC062235.1, BC088150.1, M12156.1, NM_017248.1, XM_006242384.1 transcripts.
Set Membership: |
|

Hnrnpa1
Genomic Map