Assay Details
| Gene Symbol: Zfp641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106792.1 | NP_001100262.1 | 1 - 2 | 33 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106792.1 transcripts.
Set Membership: |
|

Zfp641
Genomic Map