Assay Details
| Gene Symbol: Krt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008806.1 | NP_001008806.1 | 4 - 5 | 864 | 108 | |
| XM_006242395.2 | XP_006242457.1 | 4 - 5 | 931 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt4
Genomic Map