Assay Details
| Gene Symbol: Fkbp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013105.2 | NP_001013123.2 | 3 - 4 | 370 | 70 | |
| XM_006257344.2 | XP_006257406.1 | 2 - 3 | 1275 | 70 | ||
| XM_008765695.1 | XP_008763917.1 | 2 - 3 | 273 | 70 | ||
| XM_008765696.1 | XP_008763918.1 | 2 - 3 | 285 | 70 | ||
| XM_008765697.1 | XP_008763919.1 | 2 - 3 | 262 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008765695.1, XM_008765696.1, XM_008765697.1 transcripts.
Set Membership: |
|

Fkbp11
Genomic Map