Assay Details
| Gene Symbol: Krt71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276440.1 | NP_001263369.1 | 3 - 4 | 787 | 74 | |
| XM_008765783.1 | XP_008764005.1 | 5 - 6 | 1103 | 74 | ||
| GenBank mRNA | KF303588.1 | - | 3 - 4 | 723 | 74 | |
| KF303589.1 | - | 3 - 4 | 726 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt71
Genomic Map