Assay Details
| Gene Symbol: Krt71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276440.1 | NP_001263369.1 | 6 - 7 | 1166 | 58 | |
| XM_008765783.1 | XP_008764005.1 | 8 - 9 | 1482 | 58 | ||
| GenBank mRNA | KF303588.1 | - | 6 - 7 | 1102 | 58 | |
| KF303589.1 | - | 6 - 7 | 1105 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt71
Genomic Map