Assay Details
Gene Symbol: Krt71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276440.1 | NP_001263369.1 | 8 - 9 | 1418 | 71 | |
XM_008765783.1 | XP_008764005.1 | 10 - 11 | 1734 | 71 | ||
GenBank mRNA | KF303588.1 | - | 8 - 9 | 1354 | 71 | |
KF303589.1 | - | 8 - 9 | 1357 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
