Assay Details
| Gene Symbol: Cers5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108993.1 | NP_001102463.1 | 1 - 2 | 328 | 59 | |
| XM_006257374.2 | XP_006257436.1 | 1 - 2 | 311 | 59 | ||
| XM_006257375.2 | XP_006257437.1 | 1 - 2 | 359 | 59 | ||
| GenBank mRNA | BC166794.1 | - | 1 - 2 | 210 | 7937721 | 59 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P17622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167078 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P17622 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P17622 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P17622 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17622 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58002 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78401 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75321 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10907205 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907200 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907204 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907206 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907201 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907210 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907199 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907203 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907209 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907208 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907202 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907207 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10907198 | - | - |
| Rat Genome 230 2.0 Array | 1371676_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371676_at | - | - |
| Affymetrix HT Rat Focus Array | 1371676_at | - | - |
| Affymetrix HT Rat Focus Array | 1371676_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700031413.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031411.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031392.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031403.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031408.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031409.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031407.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031395.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031415.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031399.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031400.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031401.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031396.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031394.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031414.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031405.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031402.rn.1 | TC0700003891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700031398.rn.1 | TC0700003891.rn.1 | RTA TC |
| Rat Expression Set 230 | 1371676_at | - | - |
| Rat Expression Set 230 | 1371676_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10907198 | - | - |
| Rat Genome U34 Set | rc_AI228980_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358479 | - | - |

Cers5
Genomic Map