Assay Details
Gene Symbol: Znf740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242455.2 | XP_006242517.1 | 4 - 5 | 920 | 80 | |
XM_006242458.2 | XP_006242520.1 | 4 - 5 | 681 | 80 | ||
XM_008765870.1 | XP_008764092.1 | 4 - 5 | 717 | 80 | ||
XM_008765871.1 | XP_008764093.1 | 4 - 5 | 884 | 80 | ||
XM_008765872.1 | XP_008764094.1 | 4 - 5 | 1142 | 80 | ||
XM_008765873.1 | XP_008764095.1 | 5 - 6 | 998 | 80 | ||
XM_008765874.1 | XP_008764096.1 | 5 - 6 | 795 | 80 | ||
XM_008765875.1 | XP_008764097.1 | 3 - 4 | 567 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
