Assay Details
| Gene Symbol: Aaas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106795.1 | NP_001100265.1 | 6 - 7 | 686 | 75 | |
| XM_006242387.2 | XP_006242449.1 | 5 - 6 | 603 | 75 | ||
| XM_008765704.1 | XP_008763926.1 | 5 - 6 | 646 | 75 | ||
| XM_008765705.1 | XP_008763927.1 | 5 - 6 | 627 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aaas
Genomic Map