Assay Details
| Gene Symbol: LOC103692995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765881.1 | XP_008764103.1 | 2 - 3 | 288 | 132 | |
| Gene Symbol: Senp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765844.1 | XP_008764066.1 | 8 - 9 | 1359 | 132 | |
| XM_008765845.1 | XP_008764067.1 | 8 - 9 | 1205 | 132 | ||
| XM_008765846.1 | XP_008764068.1 | 7 - 8 | 1323 | 132 | ||
| XM_008765847.1 | XP_008764069.1 | 8 - 9 | 1359 | 132 | ||
| XM_008765848.1 | XP_008764070.1 | 8 - 9 | 832 | 132 | ||
| XM_008765849.1 | XP_008764071.1 | 7 - 8 | 1323 | 132 | ||
| XM_008765850.1 | XP_008764072.1 | 8 - 9 | 832 | 132 | ||
| GenBank mRNA | FQ230816.1 | - | 7 - 8 | 755 | 132 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103692995
Genomic Map