Assay Details
| Gene Symbol: LOC103692995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765881.1 | XP_008764103.1 | 5 - 6 | 642 | 75 | |
| Gene Symbol: Senp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765847.1 | XP_008764069.1 | 11 - 12 | 1713 | 75 | |
| XM_008765849.1 | XP_008764071.1 | 10 - 11 | 1677 | 75 | ||
| XM_008765850.1 | XP_008764072.1 | 11 - 12 | 1186 | 75 | ||
| GenBank mRNA | FQ230816.1 | - | 10 - 11 | 1109 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103692995
Genomic Map