Assay Details
| Gene Symbol: Nr4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024388.2 | NP_077364.2 | 2 - 3 | 987 | 64 | |
| XM_006242356.2 | XP_006242418.1 | 2 - 3 | 3814 | 64 | ||
| XM_006242358.2 | XP_006242420.1 | 2 - 3 | 1742 | 64 | ||
| GenBank mRNA | BC097313.1 | - | 2 - 3 | 980 | 7458540 | 64 |
| U17254.1 | - | 2 - 3 | 980 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr4a1
Genomic Map