Assay Details
Gene Symbol: Espl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170602.1 | NP_001164073.1 | 16 - 17 | 3117 | 68 | |
XM_006242460.2 | XP_006242522.1 | 16 - 17 | 3262 | 68 | ||
XM_006242461.2 | XP_006242523.1 | 16 - 17 | 3257 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
