Assay Details
Gene Symbol: Riiad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144957.1 | NP_001138429.1 | 1 - 2 | 79 | 143 | |
XM_006232847.2 | XP_006232909.1 | 1 - 2 | 339 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
