Assay Details
| Gene Symbol: Riiad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144957.1 | NP_001138429.1 | 2 - 3 | 155 | 113 | |
| GenBank mRNA | CO401253.1 | - | 2 - 3 | 119 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Riiad1
Genomic Map