Assay Details
| Gene Symbol: Rhoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106461.1 | NP_001099931.1 | 1 - 2 | 257 | 73 | |
| GenBank mRNA | BC166849.1 | - | 1 - 2 | 273 | 9088252 | 73 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC166849.1, NM_001106461.1 transcripts.
Set Membership: |
|

Rhoc
Genomic Map