Assay Details
| Gene Symbol: Rhoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233074.1 | XP_006233136.1 | 1 - 2 | 231 | 125 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006233074.1 transcripts.
Set Membership: |
|

Rhoc
Genomic Map