Assay Details
| Gene Symbol: Gpr89b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139486.1 | NP_001132958.1 | 8 - 9 | 824 | 95 | |
| XM_006232976.2 | XP_006233038.1 | 6 - 7 | 813 | 95 | ||
| GenBank mRNA | AY724489.1 | - | 8 - 9 | 755 | 95 | |
| DY546285.1 | - | 4 - 5 | 386 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr89b
Genomic Map